missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Proteinase 3/Myeloblastin/PRTN3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6134
This item is not returnable.
View return policy
Description
Proteinase 3/Myeloblastin/PRTN3 Monoclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot.
Specifications
| Proteinase 3/Myeloblastin/PRTN3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P24158 | |
| PRTN3 | |
| S. frugiperda insect ovarian cell line Sf21-derived recombinant human Proteinase 3/Myeloblastin/PRTN3 Ala26-Arg249 Accession # P24158 | |
| 100 μg | |
| Primary | |
| Detects human Proteinase 3/Myeloblastin/PRTN3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Granzyme B is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 684042 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACPA, AGP7, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, PRTN3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5657 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction