missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pref-1/DLK1/FA1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1144
This item is not returnable.
View return policy
Description
Pref-1/DLK1/FA1 Polyclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Pref-1/DLK1/FA1 | |
| Polyclonal | |
| Unconjugated | |
| AAA75364 | |
| DLK1 | |
| Mouse myeloma cell line NS0-derived recombinant human Pref-1 long isoform Ala24-Pro297 with Arg248Pro and Lys295Ser substitutions Accession # AAA75364 | |
| 100 μg | |
| Primary | |
| Detects human Pref-1/DLK-1/FA1 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Pref-1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| delta-like 1 homolog (Drosophila), DLK, DLK1, DLK-1, FA1, FA1fetal antigen 1, pG2, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8788 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction