missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PRDM16 (aa 1029-1104) Control Fragment Recombinant Protein

Product Code. 30210977
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210977

Brand: Invitrogen™ RP106278

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

PRDM16 is a zinc finger transcription factor and contains an N-terminal PR domain. The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36, q21)-positive MDS/AML. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Recent studies have shown that PRDM16 normally acts as a Smad3 binding protein that may be important for the development of orofacial structures through modulation of the TGF-beta signaling pathway. Other experiments have indicated that PRDM16 controls a bidirectional cell fate switch between skeletal myoblasts and brown fat cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HAZ2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 63976
Name Human PRDM16 (aa 1029-1104) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5730557K01Rik; CMD1LL; csp1; Histone-lysine N-methyltransferase PRDM16; Kiaa1675; line 27; LVNC8; MDS1/EVI1-like gene 1; MEL1; PFM13; PR domain 16; PR domain containing 16; PR domain zinc finger protein 16; PR domain-containing protein 16; Prdm16; Transcription factor MEL1
Common Name PRDM16
Gene Symbol PRDM16
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KHEHENAPVSQHPGVLTNHLGTSASSPTSESDNHALLDEKEDSYFSEIRNFIANSEMNQASTRTEKRADMQIVDGS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.