missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PMN (Neutrophil) Elastase ELISA Kit

Product Code. 15581857
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 15581857

Brand: Invitrogen™ BMS269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human PMN (Neutrophil) Elastase) ELISA quantitates Hu PMN (Neutrophil) Elastase in human plasma, exudate, bronchoalveolar lavage fluid, cerebrospinal fluid and seminal plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant PMN (Neutrophil) Elastase. Principle of the method The Human PMN (Neutrophil) Elastase solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08246
Assay Range 0.16 to 10.0 ng/mL
Assay Sensitivity 1.98 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cerebrospinal Fluid, Lavage Fluid, Plasma, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1991
Gene Symbol ELA2,GE,HLE,HNE,NE,PMN-E,SCN1
Interassay CV 5.6%
Intraassay CV 4.8
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cerebrospinal Fluid 10 μL, Lavage Fluid 10 μL, Plasma 10 μL, Supernatant 10 μL
Storage Requirements 2°C to 8°C
Synonym Elastase-2, leukocyte elastase,neutrophil elastase
Target Neutrophil elastase
Test Time 1 hrs 20 mins
Total Assay Time 2 hrs 20 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.