Learn More
STEMCELL Technologies Human Pluripotent Stem Cell-Derived Endoderm qPCR Array
Description
- The Human Pluripotent Stem Cell (hPSC)-Derived Endoderm Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of hPSC-derived definitive endodermal progenitor cells and their differentiated progeny including pancreatic, hepatic, and intestinal cells
- This qPCR array provides the gene expression profile of hPSC-derived endodermal cells following in vitro differentiation, and can aid in development of protocols for directed differentiation towards endodermal lineages
- Genes were selected based on their demonstrated expression in hPSCs, or in hPSC-derived ectodermal, mesodermal, and endodermal lineage cells (D'Amour et al, including pancreatic (Kroon et al, hepatic (Hay et al, and intestinal progenitor cells (Grapin-Botton & Melton)
Specifications
Specifications
| Description | Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Pancreatic Cells; Pluripotent Stem Cells |
| Format | 96-well Plate Type 3 |
| For Use With (Application) | Real Time PCR (qPCR) |