missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-1 (Research Grade Pembrolizumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
207.90 EUR - 577.50 EUR
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | Hu121 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736065
![]() |
R&D Systems
MAB11380-100 |
100 μg |
611.00 EUR 577.50 EUR / 100µg Save 33.50 EUR 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18775905
![]() |
R&D Systems
MAB11380-SP |
25 μg |
220.00 EUR 207.90 EUR / 25µg Save 12.10 EUR 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow CytometrySpecifications
| PD-1 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| 5133 | |
| IgG4 κ | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Hu121 | |
| Flow Cytometry | |
| Unconjugated | |
| Human | |
| Human | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Human PD-1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title