Learn More
Human Osteopontin/OPN Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems MAB14331-SP
 View more versions of this product
Product Code. 18730908
  • 211.00 EUR / 25µg
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Osteopontin/OPN
Monoclonal
LYOPH
Western Blot 2 ug/mL, Simple Western 100 ug/mL
NP_000573.1
SPP1
Mouse myeloma cell line NS0-derived recombinant human Osteopontin Ile17-Asn300 Accession # NP_000573.1
25 μg
Primary
Detects human Osteopontin in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse Osteopontin.
Human
Purified
Western Blot
223126
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
6696
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions