missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Obesity Panel Luminex™ Performance Control
Description
- For use as quantitative controls for the determination of human cytokine concentrations in biological fluids
- Concentrations have been assigned using R&D Systems’ Luminex™ Performance Assays
- Controls are prepared in a buffered protein solution with preservatives
- They contain recombinant human cytokines at low, medium and high concentrations
- Controls are supplied lyophilized
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Quantitative controls for the determination of human cytokine concentrations in biological fluids |
| Stability | Stable for at least 6 months from date of receipt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?