Learn More
Gibco™ Human Noggin Recombinant Protein
Description
Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in 10 mM acetic acid to a concentration of 0.1-1.0 mg/mL. Apportion the reconstituted protein into working aliquots and store at =-20°C. Make any further dilutions of the reconstituted protein in appropriate buffered solution containing a carrier protein, such as 0.1% HSA or BSA. Storage: The lyophilized protein is stable at room temperature for 3 weeks. Store at -20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at 4°C for 2-7 days, or at -20°C. Avoid repeated freeze-thaw cycles.
Specifications
Specifications
| Accession Number | Q13253 |
| For Use With (Application) | Bioactivity |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 9241 |
| Molecular Weight (g/mol) | 46.2 kDa |
| Name | Human Noggin |
| Purification Method | Purified |
| Quantity | 20 μg |
| Storage Requirements | -20°C |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.