Learn More
Invitrogen™ eBioscience™ Human NGAL Platinum ELISA Kit
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human NGAL
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
| Assay Sensitivity | 6.5pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Quantity | 96 Tests |
| Sample Volume | 50 μL |
| Synonym | Lipocalin-2, LCN2, 24p3, MSFI, NGAL, oncogene 24p3, neutrophil gelatinase-associated lipocalin |
| Target | NGAL |