missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NF-L Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 EUR - 498.00 EUR
Specifications
| Antigen | NF-L |
|---|---|
| Clone | 1033955 |
| Dilution | Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793987
![]() |
R&D Systems
MAB22164-SP |
25 μg |
201.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790867
![]() |
R&D Systems
MAB22164-100 |
100 μg |
498.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NF-L Monoclonal antibody specifically detects NF-L in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| NF-L | |
| Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 4747 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 1033955 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| 68 kDa neurofilament protein, CMT2E, NEFL, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68, NF68FLJ53642, NFL, NF-L, NFLlight polypeptide 68kDa | |
| Human NF-L peptide | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title