missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Neurotensin Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
227.00 EUR - 521.00 EUR
Specifications
| Antigen | Neurotensin |
|---|---|
| Clone | 1075611 |
| Dilution | Immunohistochemistry 3-25 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30130388
|
R&D Systems
MAB11548-100 |
100 μg |
521.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30130435
![]() |
R&D Systems
MAB11548-SP |
25 μg |
227.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurotensin Monoclonal antibody specifically detects Neurotensin in Human samples. It is validated for ImmunohistochemistrySpecifications
| Neurotensin | |
| Immunohistochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| neurotensin, neurotensin/neuromedin N, NMN-125, NN, NT, NT/N, NTS, NTS1 | |
| Neurotensin containing synthetic peptide, Accession # P30990 | |
| Primary | |
| Reconstitute lyophilized material at 0.2mg/ml in sterile PBS. For liquid material, refer to CoA for concentration. |
| 1075611 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P30990 | |
| 4922 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title