missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYOM1 Partial ORF (NP_003794.2, 1586 a.a. - 1684 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16199135
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16199135 25 μg 25µg
16189135 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16199135 Supplier Abnova™ Supplier No. H00008736Q01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

The giant protein titin, together with its associated proteins, interconnects the major structure of sarcomeres, the M bands and Z discs. The C-terminal end of the titin string extends into the M line, where it binds tightly to M-band constituents of apparent molecular masses of 190 kD (myomesin 1) and 165 kD (myomesin 2). This protein, myomesin 1, like myomesin 2, titin, and other myofibrillar proteins contains structural modules with strong homology to either fibronectin type III (motif I) or immunoglobulin C2 (motif II) domains. Myomesin 1 and myomesin 2 each have a unique N-terminal region followed by 12 modules of motif I or motif II, in the arrangement II-II-I-I-I-I-I-II-II-II-II-II. The two proteins share 50% sequence identity in this repeat-containing region. The head structure formed by these 2 proteins on one end of the titin string extends into the center of the M band. The integrating structure of the sarcomere arises from muscle-specific members of the superfamily of immunoglobulin-like proteins. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: EGKALNLTCNVWGDPPPEVSWLKNEKALASDDHCNLKFEAGRTAYFTINGVSTADSGKYGLVVKNKYGSETSDFTVSVFIPEEEARMAALESLKGGKKA

Specifications

Accession Number NP_003794.2
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8736
Molecular Weight (g/mol) 36.63kDa
Name MYOM1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen EGKALNLTCNVWGDPPPEVSWLKNEKALASDDHCNLKFEAGRTAYFTINGVSTADSGKYGLVVKNKYGSETSDFTVSVFIPEEEARMAALESLKGGKKA
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias MGC134946/MGC134947/SKELEMIN
Common Name MYOM1
Gene Symbol MYOM1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.