missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Serum Albumin Antibody, R&D Systems™

Goat Polyclonal Antibody
242.00 EUR - 653.00 EUR
Specifications
| Antigen | Albumin |
|---|---|
| Dilution | Western Blot 0.1 ug/mL, Simple Western 5-10 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755667
![]() |
R&D Systems
AF3329-SP |
25 μg |
242.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18767017
![]() |
R&D Systems
AF3329 |
100 μg |
653.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Albumin Polyclonal specifically detects Albumin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
| Albumin | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 213 | |
| Mouse Serum Albumin | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human and mouse Serum Albumin in direct ELISAs and Western blots. |
| Western Blot 0.1 ug/mL, Simple Western 5-10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Human, Mouse | |
| ALB, albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| ALB | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title