missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Growth Hormone Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 8 publications
Brand: R&D Systems AF1067-SP
This item is not returnable.
View return policy
Description
Growth Hormone Polyclonal specifically detects Growth Hormone in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Neutralization.
Specifications
| Growth Hormone | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GH1, GHB5, GHIGHD1B, GHN, GH-N, GHNGrowth hormone, GH-Nsomatotropin, growth hormone 1Pituitary growth hormone, hGH-N, IGHD1A, IGHD2, Somatotropin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2688 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 1.5-7.5 ng/mL | |
| CAA23779 | |
| GH1 | |
| E. coli-derived recombinant human Growth Hormone Phe27-Phe217 Accession # CAA23779 | |
| 25 μg | |
| Primary | |
| Detects human Growth Hormone in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction