missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech®

Product Code. 17830003
Change view
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
50µg
5µg
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17830003 250 μg 250µg
17851263 100 μg 100µg
17860103 2 x 500 μg 1mg
30089075 2 x 500 μg Pack of 2
17869882 20 μg 20µg
17801353 500 μg 500µg
17800363 50 μg 50µg
17830563 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 17830003 Supplier Gibco™ Supplier No. 12011250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

120-11-1MG will be provided as 2 x 500 μg (120-11-500UG). Recombinant Human/Murine/Rat GDF-11 is a 25.0 kDa disulfide-linked homodimer containing two 109 amino acid polypeptide chains. It is highly homologous to myostatin/GDF-8, sharing 90% amino acid sequence identity. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDF11 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95390, Q9Z1W4, Q9Z217
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 10220, 14561, 29454
Molecular Weight (g/mol) 25 kDa
Name Human/Mouse/Rat GDF-11
Purification Method Purified
Quantity 250 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BMP11; BMP-11; bone morphogenetic protein 11; GDF11; GDF-11; growth differentiation factor 11; growth/differentiation factor 11
Common Name GDF11
Gene Symbol GDF11
Biological Activity Assay 1: Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The expected ED50 for this effect is 0.08-0.10 ug/ml Assay 2: Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1cells. The ED50 for this effect is 8-10 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
Content And Storage -20°C
Activity Assay 1: Determined by its ability to inhibit induced alkaline phosphatase production by ATDC-5 chondrogenic cells. The expected ED50 for this effect is 0.08-0.10 ug/ml. Assay 2: Determined by its ability to inhibit alkaline phosphatase activity in differentiating MC3T3/E1cells. The ED50 for this effect is 8-10 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.