missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat AIF Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF1457
This item is not returnable.
View return policy
Description
AIF Polyclonal specifically detects AIF in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.
Specifications
| AIF | |
| Polyclonal | |
| Unconjugated | |
| O95831 | |
| AIFM1 | |
| E. coli-derived recombinant human AIF Glu121-Asp613 Accession # O95831 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat mitochondria-processed AIF. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| AIFapoptosis-inducing factor 1, mitochondrial, AIFM1, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9131 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction