missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FIH-1/HIF-1AN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7874
This item is not returnable.
View return policy
Description
FIH-1/HIF-1AN Monoclonal specifically detects FIH-1/HIF-1AN in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| FIH-1/HIF-1AN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1, FIH1FIH-1, FLJ20615, FLJ22027, HIF1AN, HIF-1AN, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55662 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 745122 | |
| Western Blot 0.25 ug/mL, Simple Western 12.5 ug/mL | |
| Q9NWT6 | |
| HIF1AN | |
| E. coli-derived recombinant human FIH-1/HIF-1AN Met1-Asn349 Accession # Q9NWT6 | |
| 100 μg | |
| Primary | |
| Detects human FIH-1/HIF-1AN in ELISAs. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction