missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse Doublecortin/DCX Antibody, R&D Systems™
GREENER_CHOICE

Sheep Antibody

222.00 EUR - 593.00 EUR

Specifications

Antigen Doublecortin
Dilution Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18759396
GREENER_CHOICE
View Documents
R&D Systems
AF10025-SP
25 μg
222.00 EUR
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18707516
GREENER_CHOICE
View Documents
R&D Systems
AF10025-100
100 μg
593.00 EUR
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Doublecortin specifically detects Doublecortin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Doublecortin
Western Blot, Immunohistochemistry
Unconjugated
Sheep
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
DBCN, Doublecortex, doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, LISX, X-linked (doublecortin)
E.coli-derived recombinant human Doublecortin/DCX, Met116-Thr293, Accession # O43602
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Doublecortin/DCX in direct ELISAs and Western blots. Detects mouse Doublecortin/DCX in immunohistochemistry.
Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL
Polyclonal
Lyophilized
Human, Mouse
O43602
1641
IgG
Antigen Affinity-purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.