Learn More
R&D Systems™ Human MMP Panel Luminex™ Performance Control
Product Code. 18785928
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18785928 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18785928 Supplier R&D Systems™ Supplier No. QC12B
Luminex™ bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte
Human MMP Panel Luminex™ Performance Control
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
For Use With (Equipment) Human MMP Panel
Content And Storage Store the unopened product at 2 - 8°C. Do not use past expiration date.