missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LRP-5 Intracellular Domain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6646
This item is not returnable.
View return policy
Description
LRP-5 Monoclonal specifically detects LRP-5 in Human samples. It is validated for Western Blot.
Specifications
| LRP-5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| O75197 | |
| LRP5 | |
| E. coli-derived recombinant human LRP-5 Intracellular Domain Phe1422-Ser1615 Accession # O75197 | |
| 100 μg | |
| Primary | |
| Detects human LRP-5 Intracellular Domain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with the intracellular domain of recombinant human LRP-6 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 747619 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMND1, BMND1OPTA1, EVR1, EVR4, HBM, low density lipoprotein receptor-related protein 5, low density lipoprotein receptor-related protein 7, low-density lipoprotein receptor-related protein 5, LR3VBCH2, LRP5, LRP-5, LRP7, LRP7exudative vitreoretinopathy 1, OPPG, OPS, OPTA1, osteoporosis pseudoglioma syndrome, VBCH2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4041 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction