missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Livin ELISA Kit

Product Code. 17185069
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17185069

Brand: Invitrogen™ EH302RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human Livin quantitates human Livin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Livin.

The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity. Recently, it has been identified that a novel human inhibitor of apoptosis protein (IAP) family member termed Livin or ML-IAP, which contains a single baculoviral IAP repeat (BIR) domain and a COOH-terminal RING finger domain. Livin gene has two splicing variants that contain open reading frames of 298 and 280 amino acids and both contained a single copy of baculovirus IAP repeat (BIR) and RING domain. Both of the isoforms inhibit TNFa induced apoptosis in Jurkat cells. Livin expression is low in adult tissues. It is relatively expressed at a higher level in developmental tissues and in many cancer cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number baculoviral IAP repeat containing 7; baculoviral IAP repeat-containing 7; baculoviral IAP repeat-containing 7 (livin); baculoviral IAP repeat-containing protein 7; Baculoviral IAP repeat-containing protein 7 30 kDa subunit; Baculoviral IAP repeat-containing protein 7 30 kDa subunit; Birc7; E130019N06; KIAP; Kidney inhibitor of apoptosis protein; LIVIN; livin inhibitor of apoptosis; melanoma inhibitor of apoptosis protein; MLIAP; ML-IAP; RGD1562883; RING finger protein 50; RING-type E3 ubiquitin transferase BIRC7; RNF50; RP11-261N11.7; Truncated livin; UNQ5800/PRO19607/PRO21344
Assay Range 0.1 to 25 ng/mL
Assay Sensitivity 0.1 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 79444
Gene Symbol BIRC7
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Livin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.