missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LILRA2/CD85h/ILT1 Antibody, R&D Systems™

Description
ILT1/CD85h/LILRA2 Monoclonal specifically detects ILT1/CD85h/LILRA2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | ILT1/CD85h/LILRA2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 600002 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 μg/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Accession No. | Q8N149 |
| Gene Alias | CD85 antigen-like family member H, CD85h, CD85h antigen, ILT1, ILT-1, ILT1CD85H, Leukocyte immunoglobulin-like receptor 7, leukocyte immunoglobulin-like receptor subfamily A member 2, leukocyte immunoglobulin-like receptor subfamily A member 2 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2, LIR7, LIR7LIR-7Immunoglobulin-like transcript 1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?