Learn More
Human LDLR APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 3 publications
Brand:  R&D Systems FAB2148A
Product Code. 18134421
  • 601.00 EUR / 100 tests
Description
LDLR Monoclonal specifically detects LDLR in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LDLR
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3949
Human
Purified
Flow Cytometry
472413
Flow Cytometry 10 uL/10^6 cells
P01130
LDLR
Chinese hamster ovary cell line CHO-derived recombinant human LDL R Ala22-Arg788 Accession # P01130
100 Tests
Primary
Detects human LDL R in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) LDL R, recombinant human LRP-5, or rmLRP-6 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions