missing translation for 'onlineSavingsMsg'
Learn More

Human Laforin/EPM2A Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18790908
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18790908 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18790908 Supplier R&D Systems Supplier No. MAB5714SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Laforin/EPM2A Monoclonal specifically detects Laforin/EPM2A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laforin/EPM2A
Applications Western Blot
Classification Monoclonal
Clone 523435
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAH70047
Gene Alias EC 3.1.3.16, EC 3.1.3.48, epilepsy, progressive myoclonus type 2, Lafora disease (laforin), epilepsy, progressive myoclonus type 2A, Lafora disease (laforin), EPM2, EPM2A, Lafora PTPase, laforin, LAFPTPase, LD, LDE, MELF
Gene Symbols EPM2A
Host Species Mouse
Immunogen E. coli-derived recombinant human Laforin/EPM2A Met1-Leu331 Accession # AAH70047
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7957
Test Specificity Detects human Laforin/EPM2A in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.