missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR3DL3/CD158z Antibody, R&D Systems™

Description
KIR3DL3/CD158z Monoclonal specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | KIR3DL3/CD158z |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1136B |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide |
| Gene Accession No. | Q8N743 |
| Gene Symbols | KIR3DL3 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?