Learn More
R&D Systems™ Human Kidney Biomarker Panel Luminex™ Performance Control
Description
- For use as quantitative controls for the determination of human cytokine concentrations in biological fluids.
- Concentrations have been assigned using R&D Systems Luminex™ High Performance Assays.
- Controls are prepared in a buffered protein solution with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Ea. |
| Species | Human |
| Binding Target | Luminex Assay Controls |
| For Use With (Equipment) | Human Kidney Biomarker Panel |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |