missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Kaiso (aa 113-228) Control Fragment Recombinant Protein

Product Code. 30201818
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201818

Brand: Invitrogen™ RP91977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-81890 (PA5-81890. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a transcriptional regulator with bimodal DNA-binding specificity, which binds to methylated CGCG and also to the non-methylated consensus KAISO-binding site TCCTGCNA. The protein contains an N-terminal POZ/BTB domain and 3 C-terminal zinc finger motifs. It recruits the N-CoR repressor complex to promote histone deacetylation and the formation of repressive chromatin structures in target gene promoters. It may contribute to the repression of target genes of the Wnt signaling pathway, and may also activate transcription of a subset of target genes by the recruitment of catenin delta-2 (CTNND2). Its interaction with catenin delta-1 (CTNND1) inhibits binding to both methylated and non-methylated DNA. It also interacts directly with the nuclear import receptor Importin-alpha 2 (also known as karyopherin alpha2 or RAG cohort 1), which may mediate nuclear import of this protein. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86T24
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10009
Name Human Kaiso (aa 113-228) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW260253; E130014G12Rik; Kaiso; kaiso transcription factor; RGD1566309; RP23-322E15.9; Transcriptional regulator Kaiso; WUGSC:H_DJ525N14.1; Zbtb33; zinc finger and BTB domain containing 33; zinc finger and BTB domain-containing protein 33; zinc finger transcription factor Kaiso; ZNF348; ZNF-kaiso
Common Name Kaiso
Gene Symbol ZBTB33
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IAELGVPLSQVKSISGTAQDGNTEPLPPDSGDKNLVIQKSKDEAQDNGATIMPIITESFSLSAEDYEMKKIIVTDSDDDDDDVIFCSEILPTKETLPSNNTVAQVQSNPGPVAISD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.