missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-28B/IFN-lambda 3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
219.00 EUR - 546.00 EUR
Specifications
| Antigen | IL-28B/IFN-lambda 3 |
|---|---|
| Clone | 567107 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710597
![]() |
R&D Systems
MAB52591-SP |
25 μg |
219.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18723297
![]() |
R&D Systems
MAB52591-100 |
100 μg |
546.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-28B/IFN-lambda 3 Monoclonal specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for ELISA.Specifications
| IL-28B/IFN-lambda 3 | |
| ELISA | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| interleukin-28B, IFN-lambda 3, IL28B, IL-28B, IL28C, interferon, lambda 3 | |
| Chinese hamster ovary cell line CHO-derived recombinant human IL-28B/IFN-lambda 3, Arg30-Val200 (Lys74Arg), Accession No. AAN28264 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-28B/IFN-lambda 3 in ELISAs. |
| 567107 | |
| ELISA | |
| Unconjugated | |
| Human | |
| AAN28264 | |
| 282617.0 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title