missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human HB-EGF Recombinant Protein, PeproTech®
Description
Recombinant Human HB-EGF is a 9.7 kDa protein containing 86 amino acid residues, corresponding to the extracellular EGF-like and heparin-binding domains of the full length HB-EGF protein. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
HBEGF is a growth factor that mediates its effects via EGFR, ERBB2 and ERBB4. It is required for normal cardiac valve formation and normal heart function. HBEGF promotes smooth muscle cell proliferation. It may be involved in macrophage-mediated cellular proliferation. It is mitogenic for fibroblasts, but not endothelial cells. It is able to bind EGF receptor/EGFR with higher affinity than EGF itself and is a far more potent mitogen for smooth muscle cells than EGF. It also acts as a diphtheria toxin receptor.
Specifications
Specifications
| Accession Number | Q99075 |
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 1839 |
| Molecular Weight (g/mol) | 9.7 kDa |
| Name | Human HB-EGF |
| Quantity | 500 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?