Learn More
Human Growth Hormone R/GHR Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1210-SP
This item is not returnable.
View return policy
Description
Growth Hormone R Polyclonal specifically detects Growth Hormone R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.
Specifications
| Growth Hormone R | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GH receptor, GHBP, GHR, growth hormone binding protein, growth hormone receptor, serum binding protein, Somatotropin receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2690 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Neutralization 0.125-0.5 ug/mL, CyTOF-ready | |
| P10912 | |
| GHR | |
| Mouse myeloma cell line NS0-derived recombinant human Growth Hormone R/GHR Ala27-Tyr264 Accession # P10912 | |
| 25 μg | |
| Primary | |
| Detects human Growth Hormone R/GHR in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant mouse GHR and recombinant rat GHR is observed. | |
| Human | |
| IgG |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction