Learn More
Human GIP Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8864
This item is not returnable.
View return policy
Description
GIP Monoclonal specifically detects GIP in Human samples. It is validated for Immunohistochemistry.
Specifications
| GIP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide, Incretin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2695 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 911205 | |
| Immunohistochemistry 8-25 ug/mL | |
| P09681 | |
| GIP | |
| GIP Peptide Tyr1-Asn39 Accession # P09681 | |
| 100 μg | |
| Primary | |
| Detects human GIP in direct ELISAs. | |
| Human | |
| Supernatant |