Learn More
Human GIP Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB8864
Product Code. 18752718
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
GIP Monoclonal specifically detects GIP in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GIP
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
gastric inhibitory polypeptide, Glucose-dependent insulinotropic polypeptide, Incretin
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2695
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Immunohistochemistry
911205
Immunohistochemistry 8-25 ug/mL
P09681
GIP
GIP Peptide Tyr1-Asn39 Accession # P09681
100 μg
Primary
Detects human GIP in direct ELISAs.
Human
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions