missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GDF-8/Myostatin ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human GDF-8/Myostatin quantitates human GDF-8/Myostatin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human GDF-8/Myostatin.
The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
Specifications
Specifications
| Accession Number | Cmpt; compact; GDF8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; myostatin |
| Assay Range | 0.83 to 200 ng/mL |
| Assay Sensitivity | 0.83 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2660 |
| Gene Symbol | Mstn |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction