missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Gastrin ELISA Kit

Product Code. 18007404
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18007404 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18007404 Supplier Invitrogen™ Supplier No. EEL058

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

This antibody may be used to study the differential tissue expression and intracellular and subcellular localization of gastrin in neuroendocrine cells of the gastrointestinal tract, and in the CNS. Antibodies to gastrin are also useful for the identification and detection of gastrin in normal and neoplastic tissue.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01350
Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2520
Gene Symbol Gast
Interassay CV <10%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Gastrin
Test Time 1 hrs 20 mins
Total Assay Time 3 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.