missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Gas6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
208.00 EUR - 2030.00 EUR
Specifications
| Antigen | Gas6 |
|---|---|
| Clone | 100127 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728389
![]() |
R&D Systems
MAB885-SP |
25 μg |
208.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759937
![]() |
R&D Systems
MAB885-100 |
100 μg |
523.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754299
![]() |
R&D Systems
MAB885-500 |
500 μg |
2030.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gas6 Monoclonal specifically detects Gas6 in Human samples. It is validated for Western Blot.Specifications
| Gas6 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| NP_000811 | |
| 2621 | |
| Mouse myeloma cell line NS0-derived recombinant human Gas6 Asp118-Ala678 Accession # NP_000811 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Gas6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 5% cross-reactivity with recombinant mouse Gas6. |
| 100127 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| GAS6 | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title