missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Galectin-8 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1305
This item is not returnable.
View return policy
Description
Galectin-8 Polyclonal specifically detects Galectin-8 in Human samples. It is validated for Western Blot.
Specifications
| Galectin-8 | |
| Polyclonal | |
| Biotin | |
| O00214 | |
| LGALS8 | |
| E. coli-derived recombinant human Galectin-8 Met1-Trp317 Accession # O00214 | |
| 50 μg | |
| Primary | |
| Detects human Galectin-8 in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human (rh) Galectin-1, rhGalectin-2, rhGalectin-3, rhGalectin-4, and rhGalectin-7 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Gal-8, GAL8, galectin 8, galectin-8, Galectin8, galectin-8g, lectin, galactoside-binding, soluble, 8, LGALS8, PCTA1, PCTA-1Po66 carbohydrate-binding protein, Po66 carbohydrate binding protein, Po66-CBP, Prostate carcinoma tumor antigen 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3964 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction