missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FUT10 (aa 366-435) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63601 (PA5-63601. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Probable fucosyltransferase.
Specifications
Specifications
| Accession Number | Q6P4F1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 84750 |
| Name | Human FUT10 (aa 366-435) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | alpha (1,3) fucosyltransferase; alpha 1,3-fucosyl transferase; alpha-(1,3)-fucosyltransferase 10; alpha3-fucosyltransferase; Fucosyltransferase 10; fucosyltransferase 10 (alpha (1,3) fucosyltransferase); fucosyltransferase X; FUCTX; fuc-TX; fucT-X; FUT10; fut10A; Galactoside 3-L-fucosyltransferase 10 |
| Common Name | FUT10 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction