missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fc gamma RIIB/C (CD32b/c) Antibody, R&D Systems™

Description
Fc gamma RIIB/C (CD32b/c) Monoclonal specifically detects Fc gamma RIIB/C (CD32b/c) in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
| Antigen | Fc gamma RIIB/C (CD32b/c) |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 190710 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P31994 |
| Gene Symbols | FCGR2B |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?