missing translation for 'onlineSavingsMsg'
Learn More

Human Fc gamma RII/CD32 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18008971
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18008971 25 μg 25µg
18029771 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18008971 Supplier R&D Systems Supplier No. AF1875SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Fc gamma RII/CD32 Polyclonal antibody specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fc gamma RII/CD32
Applications Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA35827
Gene Alias CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, FcgRII, FcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b
Gene Symbols FCGR2A
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Fc gamma RIIA/CD32a Ala36-Ile218 Accession # AAA35827
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2213
Test Specificity Detects human Fc gamma RII/CD32 in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Fc gamma RIIIB is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.