Learn More
Human Fc gamma RI/CD64 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
213.00 EUR - 2040.00 EUR
Specifications
| Antigen | Fc gamma RI/CD64 |
|---|---|
| Clone | 10.1 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710767
![]() |
R&D Systems
MAB1257-SP |
25 μg |
213.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797178
![]() |
R&D Systems
MAB1257-100 |
100 μg |
517.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742939
![]() |
R&D Systems
MAB1257-500 |
500 μg |
2040.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with rheumatoid synovial fluid cells and fibronectin purified monocytes. The IgG fraction of the tissue culture supernatant was purified by Protein G affinity chromatography.
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| Fc gamma RI/CD64 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| FCGR1A | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 10.1 | |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2209 | |
| Rheumatoid synovial fluid cells and fibronectin purified monocytes | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Fc gamma RI/CD64. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.