missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ER alpha/NR3A1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF5715
This item is not returnable.
View return policy
Description
ER alpha/NR3A1 Polyclonal specifically detects ER alpha/NR3A1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| ER alpha/NR3A1 | |
| Polyclonal | |
| Unconjugated | |
| P03372 | |
| ESR1 | |
| E. coli-derived recombinant human ER alpha Met1-Gln116 Accession # P03372 | |
| 100 μg | |
| Primary | |
| Detects human ER alpha . | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ER, ER alpha, Era, ER-alpha, ESR1, ESRESRA, Estradiol receptor, estrogen receptor, estrogen receptor 1, estrogen receptor alpha, estrogen receptor alpha delta 3*4,56,7*/819-2 isoform, estrogen receptor alpha delta 4 +49 isoform, estrogen receptor alpha delta 4*5,6,7*/654 isoform, NR3A1, NR3A1DKFZp686N23123, Nuclear receptor subfamily 3 group A member 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2099 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction