missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EphB3 ELISA Kit

Product Code. 17124929
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17124929

Brand: Invitrogen™ EH174RBX5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human EphB3 quantitates human EphB3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human EphB3.

EPHB3 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. The ephrin ligands and Eph receptors display reciprocal expression in vivo. Developing and adult neural tissue express nearly all of the Eph receptors and ephrin ligands. Ephs and ephrins play a significant role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AW456895; CAPB; Cek10; Developmental kinase 5; DRT; EK2; ELK; Embryonic kinase 2; EPH receptor B3; EPHB2; Ephb3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin B1; Ephrin B2; Ephrin B3; ephrin type-B receptor 3; EphrinB1; EphrinB2; EphrinB3; EPHT3; ERK; Etk2; HEK2; Hek5; HEK-6; human embryo kinase 2; Mdk5; mDK-5; MGC87492; NET; PCBC; Sek4; Tyro5; TYRO6; Tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6
Assay Range 1.8 to 400 ng/mL
Assay Sensitivity 1.8 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2049
Gene Symbol EPHB3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target EphB3
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.