missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EDNRA Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB6538
This item is not returnable.
View return policy
Description
EDNRA/Endothelin R Type A Monoclonal specifically detects EDNRA/Endothelin R Type A in Human samples. It is validated for Immunohistochemistry.
Specifications
| EDNRA/Endothelin R Type A | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| NP_001948 | |
| EDNRA | |
| NS0 mouse myeloma cell line transfected with human EDRNA Accession # NP_001948 | |
| 100 μg | |
| Primary | |
| Detects human EDRNA in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 485711 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EDNRA, endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1909 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction