missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNAH5 (aa 921-1008) Control Fragment Recombinant Protein

Código de producto. 30207085
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30207085

missing translation for 'mfr': Invitrogen™ RP96795

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57957 (PA5-57957. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a dynein protein, which is part of a microtubule-associated motor protein complex consisting of heavy, light, and intermediate chains. This protein is an axonemal heavy chain dynein. It functions as a force-generating protein with ATPase activity, whereby the release of ADP is thought to produce the force-producing power stroke. Mutations in this gene cause primary ciliary dyskinesia type 3, as well as Kartagener syndrome, which are both diseases due to ciliary defects.
TRUSTED_SUSTAINABILITY

Especificaciones

Accession Number Q8TE73
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1767
Name Human DNAH5 (aa 921-1008) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AU022615; Axonemal beta dynein heavy chain 5; axonemal dynein heavy chain 5; b2b1134Clo; b2b1154Clo; b2b1537Clo; b2b1565Clo; b2b601Clo; CILD3; Ciliary dynein heavy chain 5; DLP5; Dnah5; Dnahc5; Dnahc8; dynein axonemal heavy chain 5; dynein heavy chain 5, axonemal; dynein, axon, heavy chain 5; dynein, axonemal, heavy chain 5; dynein, axonemal, heavy polypeptide 5; dynein-like protein 5; HL1; KIAA1603; KTGNR; Mdnah5; mKIAA1603; PCD
Common Name DNAH5
Gene Symbol DNAH5
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NFDTLTSSINARANALLLTTVTRKKKETEMLGEEARELLSHFNHQNMDALLKVTRNTLEAIRKRIHSSHTINFRDSNSASNMKQNSLP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado