missing translation for 'onlineSavingsMsg'
Learn More

Human DGK-beta Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18795219
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18795219 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18795219 Lieferant R&D Systems Lieferanten-Nr. MAB6727SP

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

DGK-beta Monoclonal specifically detects DGK-beta in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen DGK-beta
Applications Western Blot
Classification Monoclonal
Clone 646523
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y6T7
Gene Alias 90 kDa diacylglycerol kinase, DAG kinase beta, DAGK2, DGKB, DGKbeta, DGK-beta, diacylglycerol kinase beta, diacylglycerol kinase, beta (90kD), diacylglycerol kinase, beta 90kDa, Diglyceride kinase beta, EC 2.7.1, EC 2.7.1.107, KIAA0718DGKDGK-BETA
Gene Symbols DGKB
Host Species Mouse
Immunogen E. coli-derived recombinant human DGK-beta Met1-Ser143 Accession # Q9Y6T7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1607
Test Specificity Detects human DGK-beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human DGK-alpha , gamma , epsilon , iota , kappa , theta , or zeta is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Mehr anzeigen Weniger anzeigen

For Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.