Learn More
Human Cystatin C Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11963-100
 View more versions of this product
Product Code. 18701497
  • 582.00 EUR / 100µg
Description
Cystatin C Monoclonal antibody specifically detects Cystatin C in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin C
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin
Mouse myeloma cell line NS0-derived recombinant human Cystatin C, Ser27-Ala146, Accession # P01034
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2b
Immunohistochemistry, Immunocytochemistry
197814
Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
P01034
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
1471
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions