missing translation for 'onlineSavingsMsg'
Learn More

Human CTRP9/C1qTNF9 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18742689
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18742689 25 μg 25µg
18121961 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18742689 Supplier R&D Systems Supplier No. MAB6537SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTRP9/C1qTNF9 Monoclonal specifically detects CTRP9/C1qTNF9 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CTRP9/C1qTNF9
Applications Western Blot
Classification Monoclonal
Clone 653533
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P0C862
Gene Alias AQL1, C1q and tumor necrosis factor related protein 9, C1qTNF9, C1QTNF9AComplement C1q tumor necrosis factor-related protein 9, complement C1q tumor necrosis factor-related protein 9A, CTRP9, MGC48915
Gene Symbols C1QTNF9
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CTRP9/C1qTNF9 Asn16-Pro333 Accession # P0C862
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338872
Test Specificity Detects human CTRP9/C1qTNF9 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1qTNF1, rhCORS26/C1qTNF3, rhC1qTNF4, rhC1qTNF5, rhC1QL2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.