missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Spike S1 Rabbit anti-HCoV-HKU1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
802.00 EUR - 2440.00 EUR
Specifications
| Antigen | Human Coronavirus Spike S1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18342322
|
Novus Biologicals
NBP3-14865-0.1MG |
0.1 mg |
802.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18399421
|
Novus Biologicals
NBP3-14865-0.5MG |
0.5 mg |
2440.00 EUR
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human Coronavirus Spike S1 Polyclonal antibody specifically detects Human Coronavirus Spike S1 in HCoV-HKU1 samples. It is validated for ELISASpecifications
| Human Coronavirus Spike S1 | |
| ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 0.2 um filtered. | |
| IgG | |
| Antigen Affinity-purified | |
| Antibody |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| HCoV-HKU1 | |
| Recombinant HCoV-HKU1 spike subunit 1 (S1) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title