missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human cIAP-1/BIRC2/hIAP-2 ELISA Kit

Product Code. 17154383
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
Pack of 96
This item is not returnable. View return policy

Product Code. 17154383

Brand: Invitrogen™ EH108RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Human cIAP-1/BIRC2/hIAP-2 quantitates human cIAP-1/BIRC2/hIAP-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human cIAP-1/BIRC2/hIAP-2.

The protein encoded by this gene is a member of a family of proteins that inhibits apoptosis by binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2, probably by interfering with activation of ICE-like proteases. This encoded protein inhibits apoptosis induced by serum deprivation and menadione, a potent inducer of free radicals.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number API1; Api2; apoptosis inhibitor 1; apoptosis inhibitor 2; AW146227; baculoviral IAP repeat containing 2; baculoviral IAP repeat-containing 2; baculoviral IAP repeat-containing 3; baculoviral IAP repeat-containing protein 2; BIRC2; Birc3; Cellular inhibitor of apoptosis 1; cIAP1; c-IAP1; cIAP2; HIAP1; HIAP2; Hiap-2; IAP homolog B; IAP1; IAP2; IAP-2; inhibitor of apoptosis protein; inhibitor of apoptosis protein 2; mcIAP1; MIAP1; MIAP2; mIAP-2; MIHB; MIHC; NFR2-TRAF signalling complex protein; rIAP1; RING finger protein 48; RING-type E3 ubiquitin transferase BIRC2; RNF48; TNFR2-TRAF-signaling complex protein 2
Assay Range 1.65 to 400 ng/mL
Assay Sensitivity 1.65 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 329
Gene Symbol BIRC2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target cIAP1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.