missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CDC42EP2 Full-length ORF (NP_006770.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Product Code. 16125082
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16125082 10 μg 10µg
16135082 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16125082 Supplier Abnova™ Supplier No. H00010435P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for AP, Array, ELISA, WB-Re

CDC42, a small Rho GTPase, regulates the formation of F-actin-containing structures through its interaction with the downstream effector proteins. The protein encoded by this gene is a member of the Borg family of CDC42 effector proteins. Borg family proteins contain a CRIB (Cdc42/Rac interactive-binding) domain. They bind to, and negatively regulate the function of, CDC42. Coexpression of this protein with dominant negative mutant CDC42 protein in fibroblast was found to induce pseudopodia formation, which suggested a role of this protein in actin filament assembly and cell shape control. [provided by RefSeq]

Sequence: MSTKVPIYLKRGSRKGKKEKLRDLLSSDMISPPLGDFRHTIHIGSGGGSDMFGDISFLQGKFHLLPGTMVEGPEEDGTFDLPFQFTRTATVCGRELPDGPSPLLKNAISLPVIGGPQALTLPTAQAPPKPPRLHLETPQPSPQEGGSVDIWRIPETGSPNSGLTPESGAEEPFLSNASSLLSLHVDLGPSILDDVLQIMDQDLDSMQIPT

Specifications

Accession Number NP_006770.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10435
Molecular Weight (g/mol) 48.9kDa
Name CDC42EP2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MSTKVPIYLKRGSRKGKKEKLRDLLSSDMISPPLGDFRHTIHIGSGGGSDMFGDISFLQGKFHLLPGTMVEGPEEDGTFDLPFQFTRTATVCGRELPDGPSPLLKNAISLPVIGGPQALTLPTAQAPPKPPRLHLETPQPSPQEGGSVDIWRIPETGSPNSGLTPESGAEEPFLSNASSLLSLHVDLGPSILDDVLQIMDQDLDSMQIPT
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias BORG1/CEP2
Common Name CDC42EP2
Gene Symbol CDC42EP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.